ENA245487727 Enamine

1-(2-chlorophenyl)ethane-1-thiol

  • MDL number MFCD09809852
  • Cas Number : 1016716-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55341
Availability details for
Quantity
  • Availability
  • Add to Cart