ENA245487734 Enamine

ethyl 2-(tert-butylamino)acetate

  • MDL number MFCD00239220
  • Cas Number : 37885-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55764
Availability details for
Quantity
  • Availability
  • Add to Cart