ENA245487737 Enamine

3-methyl-7-propyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione

  • MDL number MFCD02089337
  • Cas Number : 55242-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55770
Availability details for
Quantity
  • Availability
  • Add to Cart