ENA245487742 Enamine

5-methyl-3-(3-nitrophenyl)-1,2,4-oxadiazole

  • MDL number MFCD00740195
  • Cas Number : 25283-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55775

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55775
Availability details for
Quantity
  • Availability
  • Add to Cart