ENA245487745 Enamine

3-[(2-carbamoylphenoxy)methyl]benzoic acid

  • MDL number MFCD09929341
  • Cas Number : 891831-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55778
Availability details for
Quantity
  • Availability
  • Add to Cart