ENA245487747 Enamine

3-(aminomethyl)-N-[(oxolan-2-yl)methyl]benzamide

  • MDL number MFCD09807956
  • Cas Number : 1016527-39-1
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55344
Availability details for
Quantity
  • Availability
  • Add to Cart