ENA245487748 Enamine

2-(2,5-dioxopyrrolidin-1-yl)-2-phenylacetic acid

  • MDL number MFCD09729608
  • Cas Number : 1500-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55783
Availability details for
Quantity
  • Availability
  • Add to Cart