ENA245487753 Enamine

2-(4-chloro-2-nitrophenoxy)propanoic acid

  • MDL number MFCD03422225
  • Cas Number : 21105-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55789
Availability details for
Quantity
  • Availability
  • Add to Cart