ENA245487755 Enamine

1-methyl-1,2,3,4-tetrahydroquinolin-6-amine

  • MDL number MFCD09035177
  • Cas Number : 91817-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55792

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55792
Availability details for
Quantity
  • Availability
  • Add to Cart