ENA245487756 Enamine

[1-(2-methoxyethyl)-3,5-dimethyl-1H-pyrazol-4-yl]methanamine

  • MDL number MFCD12103783
  • Cas Number : 1155595-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55793
Availability details for
Quantity
  • Availability
  • Add to Cart