ENA245487762 Enamine

9-benzyl-9-azabicyclo[3.3.1]nonan-3-amine

  • MDL number MFCD13806395
  • Cas Number : 76272-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55802
Availability details for
Quantity
  • Availability
  • Add to Cart