ENA245487769 Enamine

5-methoxy-2-(pyridin-2-ylmethoxy)benzaldehyde

  • MDL number MFCD08442767
  • Cas Number : 926240-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55273

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55273
Availability details for
Quantity
  • Availability
  • Add to Cart