ENA245487770 Enamine

3-[(4-benzylpiperazin-1-yl)sulfonyl]-4-fluorobenzoic acid

  • MDL number MFCD13196284
  • Cas Number : 1054112-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55816
Availability details for
Quantity
  • Availability
  • Add to Cart