ENA245487771 Enamine

3-{4-[(4-ethylpiperazin-1-yl)sulfonyl]phenyl}propanoic acid

  • MDL number MFCD13196285
  • Cas Number : 1044511-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55817
Availability details for
Quantity
  • Availability
  • Add to Cart