ENA245487772 Enamine

3-[4-(2-methylpropoxy)phenyl]-1-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD02705588
  • Cas Number : 371776-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55818

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55818
Availability details for
Quantity
  • Availability
  • Add to Cart