ENA245487793 Enamine

2-{[(thiophen-3-yl)methyl]sulfanyl}ethan-1-amine

  • MDL number MFCD07366437
  • Cas Number : 901273-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55842
Availability details for
Quantity
  • Availability
  • Add to Cart