ENA245487810 Enamine

3-amino-1-butylurea

  • MDL number MFCD00047818
  • Cas Number : 20605-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55865
Availability details for
Quantity
  • Availability
  • Add to Cart