ENA245487813 Enamine

methyl 2-[(4-bromobenzene)sulfonyl]acetate

  • MDL number MFCD11642539
  • Cas Number : 50397-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55868

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55868
Availability details for
Quantity
  • Availability
  • Add to Cart