ENA245487834 Enamine

2-(difluoromethoxy)-5-nitrobenzaldehyde

  • MDL number MFCD00521794
  • Cas Number : 145742-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55904
Availability details for
Quantity
  • Availability
  • Add to Cart