ENA245487837 Enamine

2-(carbamoylamino)-4-chlorobenzoic acid

  • MDL number MFCD09048937
  • Cas Number : 214288-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55907
Availability details for
Quantity
  • Availability
  • Add to Cart