ENA245487839 Enamine

2-[(1,3-thiazol-2-yl)amino]acetic acid

  • MDL number MFCD00458149
  • Cas Number : 79858-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55372
Availability details for
Quantity
  • Availability
  • Add to Cart