ENA245487844 Enamine

2-ethyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD01760299
  • Cas Number : 30709-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-157
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55918
Availability details for
Quantity
  • Availability
  • Add to Cart