ENA245487847 Enamine

6-cyclopropyl-1-(1-cyclopropylethyl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09738364
  • Cas Number : 954254-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55921
Availability details for
Quantity
  • Availability
  • Add to Cart