ENA245487852 Enamine

oxolane-2-carbonyl chloride

  • MDL number MFCD09897727
  • Cas Number : 52449-98-6
  • Molecular Weight : 134.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55255

Safety Information

signalword Danger
pcodes P210,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-55255
pictograms GHS05
hstatements H227,H314
Availability details for
Quantity
  • Availability
  • Add to Cart