ENA245487864 Enamine

4-{5-bromo-1H-pyrazolo[3,4-b]pyridin-1-yl}-1-methylpiperidine

  • MDL number MFCD09946632
  • Cas Number : 1016850-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120...122
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55956

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55956
Availability details for
Quantity
  • Availability
  • Add to Cart