ENA245487865 Enamine

2-(trifluoromethyl)quinolin-7-ol

  • MDL number MFCD02192593
  • Cas Number : 41192-81-8
  • Molecular Weight : 213.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55388
Availability details for
Quantity
  • Availability
  • Add to Cart