ENA245487869 Enamine

4-(aminomethyl)-N-(pyridin-4-yl)benzamide dihydrochloride

  • MDL number MFCD13196312
  • Cas Number : 173897-49-9
  • Molecular Weight : 300.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55962
Availability details for
Quantity
  • Availability
  • Add to Cart