ENA245487874 Enamine

(4-fluorophenyl)methanesulfonohydrazide

  • MDL number MFCD12181287
  • Cas Number : 1155081-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55968
Availability details for
Quantity
  • Availability
  • Add to Cart