ENA245487875 Enamine

2-[(2-chlorophenyl)methoxy]ethan-1-amine hydrochloride

  • MDL number MFCD01626089
  • Cas Number : 6594-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55969
Availability details for
Quantity
  • Availability
  • Add to Cart