ENA245487877 Enamine

1-(4-methyl-1,3-thiazol-2-yl)piperidine-4-carboxylic acid

  • MDL number MFCD09735733
  • Cas Number : 953720-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55971
Availability details for
Quantity
  • Availability
  • Add to Cart