ENA245487882 Enamine

4-(4-bromo-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD06167046
  • Cas Number : 898054-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55284

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55284
Availability details for
Quantity
  • Availability
  • Add to Cart