ENA245487883 Enamine

3-[4-(2-methylpropoxy)phenyl]-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04639701
  • Cas Number : 955972-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55980
Availability details for
Quantity
  • Availability
  • Add to Cart