ENA245487886 Enamine

3-(3-nitrophenyl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD02708777
  • Cas Number : 376358-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55984
Availability details for
Quantity
  • Availability
  • Add to Cart