ENA245487890 Enamine

1-phenyl-3-(pyridin-4-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD05987121
  • Cas Number : 957354-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55988
Availability details for
Quantity
  • Availability
  • Add to Cart