ENA245487891 Enamine

3-(5-chlorothiophen-2-yl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04623019
  • Cas Number : 882228-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55989
Availability details for
Quantity
  • Availability
  • Add to Cart