ENA245487892 Enamine

1-(2-cyanoethyl)-3-(3-nitrophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06359991
  • Cas Number : 956984-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55991
Availability details for
Quantity
  • Availability
  • Add to Cart