ENA245487894 Enamine

1-(2-cyanoethyl)-3-(3,4-dimethylphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06359994
  • Cas Number : 956411-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55993
Availability details for
Quantity
  • Availability
  • Add to Cart