ENA245487898 Enamine

3-(2,2-dimethylpropyl)-6-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09740553
  • Cas Number : 954577-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55998
Availability details for
Quantity
  • Availability
  • Add to Cart