ENA245487899 Enamine

1-cyclohexyl-6-cyclopropyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09730347
  • Cas Number : 953891-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55999
Availability details for
Quantity
  • Availability
  • Add to Cart