ENA245487900 Enamine

3-(furan-2-yl)-6-methyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09738365
  • Cas Number : 954254-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56000
Availability details for
Quantity
  • Availability
  • Add to Cart