ENA245487905 Enamine

3-fluoro-4-[2-(trifluoromethyl)phenoxy]aniline

  • MDL number MFCD12469916
  • Cas Number : 1039868-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56006

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56006
Availability details for
Quantity
  • Availability
  • Add to Cart