ENA245487931 Enamine

4-phenoxybutanimidamide hydrochloride

  • MDL number MFCD09260747
  • Cas Number : 1071580-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55263
Availability details for
Quantity
  • Availability
  • Add to Cart