ENA245487935 Enamine

2-(2-bromophenyl)-4-(chloromethyl)-1,3-oxazole

  • MDL number MFCD11200992
  • Cas Number : 1038295-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56080
Availability details for
Quantity
  • Availability
  • Add to Cart