ENA245487937 Enamine

1-(4-methyl-1,3-thiazol-2-yl)piperidine-3-carboxylic acid

  • MDL number MFCD09054881
  • Cas Number : 927803-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56082
Availability details for
Quantity
  • Availability
  • Add to Cart