ENA245487941 Enamine

3-[(octyloxy)methyl]aniline

  • MDL number MFCD11196400
  • Cas Number : 1039933-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56089
Availability details for
Quantity
  • Availability
  • Add to Cart