ENA245487942 Enamine

6-methoxy-N-methylpyridin-3-amine

  • MDL number MFCD10697832
  • Cas Number : 61771-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56091
Availability details for
Quantity
  • Availability
  • Add to Cart