ENA245487945 Enamine

tert-butyl 3,7-diazabicyclo[4.2.0]octane-3-carboxylate

  • MDL number MFCD08234914
  • Cas Number : 885271-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56100
Availability details for
Quantity
  • Availability
  • Add to Cart