ENA245487947 Enamine

1-(2-cyanoethyl)-3-(naphthalen-1-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD07392146
  • Cas Number : 957369-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56104
Availability details for
Quantity
  • Availability
  • Add to Cart