ENA245487988 Enamine

2-[(4-methylpiperazin-1-yl)methyl]-1H-1,3-benzodiazol-6-amine

  • MDL number MFCD11978899
  • Cas Number : 1096321-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55290
Availability details for
Quantity
  • Availability
  • Add to Cart