ENA245487995 Enamine

1-amino-3-(2,3-dichlorophenoxy)propan-2-ol

  • MDL number MFCD09950348
  • Cas Number : 23105-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55291
Availability details for
Quantity
  • Availability
  • Add to Cart